APO455827928 Apollo Scientific Ltd

4-Bromo-1H-indole, N-BOC protected

  • MDL number MFCD04117874
  • Cas Number : 676448-17-2
  • Molecular Weight : 296.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-bromo-1H-indole-1-carboxylate
Inchi Code 1S/C13H14BrNO2/c1-13(2,3)17-12(16)15-8-7-9-10(14)5-4-6-11(9)15/h4-8H,1-3H3
Inchi Key ZGBNKNOAADXGOH-UHFFFAOYSA-N

Description

Synonym tert-Butyl 4-bromo-1H-indole-1-carboxylate, 4-Bromo-1-(tert-butoxycarbonyl)-1H-indole
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1h-indole-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1h-indole-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart