APO455827936 Apollo Scientific Ltd

3-Bromo-5-chloropyridine-2-carboxylic acid

  • MDL number MFCD12827551
  • Cas Number : 1189513-50-5
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-chloro-2-pyridinecarboxylic acid
Inchi Code 1S/C6H3BrClNO2/c7-4-1-3(8)2-9-5(4)6(10)11/h1-2H,(H,10,11)
Inchi Key IUWXGYGVXBPNHT-UHFFFAOYSA-N

Description

Synonym 3-Bromo-2-carboxy-5-chloropyridine, 3-Bromo-5-chloropicolinic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-chloropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-chloropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart