APO455827941 Apollo Scientific Ltd

5-[(Morpholin-4-yl)methyl]isoxazole-3-carboxylic acid hydrochloride

  • MDL number MFCD08277061
  • Cas Number : 944450-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-160
Synonym 5-[(Morpholin-4-yl)methyl]-1,2-oxazole-3-carboxylic acid hydrochloride, 4-[(3-Carboxyisoxazol-5-yl)methyl]morpholine hydrochloride
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46277

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46277
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart