APO455827945 Apollo Scientific Ltd

4-Amino-2-methoxypyrimidine

  • MDL number MFCD00128201
  • Cas Number : 3289-47-2
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methoxy-4-pyrimidinylamine
Inchi Code 1S/C5H7N3O/c1-9-5-7-3-2-4(6)8-5/h2-3H,1H3,(H2,6,7,8)
Inchi Key DHYLZDVDOQLEAQ-UHFFFAOYSA-N

Description

Synonym 2-Methoxypyrimidin-4-amine, 4-Amino-2-methoxy-1,3-diazine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-2-methoxypyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-2-methoxypyrimidine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart