APO455827948 Apollo Scientific Ltd

4-Sulphamoylbenzeneboronic acid, pinacol ester

  • MDL number MFCD06657806
  • Cas Number : 214360-51-7
  • Molecular Weight : 283.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide
Purity 98
Storage temp. Store at 2-8
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-sulphamoylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-sulphamoylbenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart