APO455827951 Apollo Scientific Ltd

5-Bromobenzo[c]isothiazole

  • MDL number MFCD08706414
  • Cas Number : 20712-07-6
  • Molecular Weight : 214.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,1-benzisothiazole
Inchi Code 1S/C7H4BrNS/c8-6-1-2-7-5(3-6)4-10-9-7/h1-4H
Inchi Key OZRILMBPXSZJAR-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2,1-benzothiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromobenzocisothiazole

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/5-bromobenzocisothiazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart