APO455827961 Apollo Scientific Ltd

5-Iodo-1H-benzimidazole

  • MDL number MFCD06659631
  • Cas Number : 78597-27-0
  • Molecular Weight : 244.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-145
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-iodo-1h-benzimidazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-iodo-1h-benzimidazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart