APO455827963 Apollo Scientific Ltd

3-Methyl-2-nitrobenzaldehyde

  • MDL number MFCD08703364
  • Cas Number : 5858-27-5
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 59-61
Iupac Name 3-methyl-2-nitrobenzaldehyde
Inchi Code 1S/C8H7NO3/c1-6-3-2-4-7(5-10)8(6)9(11)12/h2-5H,1H3
Inchi Key HAGUMWGXABFJMN-UHFFFAOYSA-N

Description

Synonym 2-Nitro-m-tolualdehyde, 3-Formyl-2-nitrotoluene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 59-61
Product Link https://store.apolloscientific.co.uk/product/3-methyl-2-nitrobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-2-nitrobenzaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart