APO455827964 Apollo Scientific Ltd

7-Nitro-1H-indole-3-carboxaldehyde

  • MDL number MFCD01074519
  • Cas Number : 10553-14-7
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-nitro-1H-indole-3-carbaldehyde
Inchi Code 1S/C9H6N2O3/c12-5-6-4-10-9-7(6)2-1-3-8(9)11(13)14/h1-5,10H
Inchi Key ADGKBVRTGVODMM-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-7-nitro-1H-indole
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-nitro-1h-indole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/7-nitro-1h-indole-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart