APO455827966 Apollo Scientific Ltd

5-(Benzyloxy)-3-formyl-1H-indole, N-BOC protected

  • MDL number MFCD05864701
  • Cas Number : 914348-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Benzyloxy)-1-(tert-butoxycarbonyl)-1H-indole-3-carboxaldehyde, tert-Butyl 5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate
Purity 98
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29679

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29679
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart