APO455827970 Apollo Scientific Ltd

4-Chloro-3-formyl-1H-indole, N-BOC protected

  • MDL number MFCD05864705
  • Cas Number : 914349-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 4-chloro-3-formyl-1H-indole-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-chloro-1H-indole-3-carboxaldehyde
Purity 98
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29687

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29687
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart