APO455827974 Apollo Scientific Ltd

5-Chloro-2-(hydroxymethyl)pyridine

  • MDL number MFCD10000856
  • Cas Number : 209526-98-7
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 45-47
Iupac Name (5-chloro-2-pyridinyl)methanol
Inchi Code 1S/C6H6ClNO/c7-5-1-2-6(4-9)8-3-5/h1-3,9H,4H2
Inchi Key JJNKKECPOPWYNY-UHFFFAOYSA-N

Description

Synonym (5-Chloropyridin-2-yl)methanol, 5-Chloropicolinyl alcohol
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 45-47
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-hydroxymethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-hydroxymethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart