APO455827976 Apollo Scientific Ltd

Methyl 3-bromopyridine-2-carboxylate

  • MDL number MFCD12025946
  • Cas Number : 53636-56-9
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-bromopicolinate, 3-Bromo-2-(methoxycarbonyl)pyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromopyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P260;P280;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromopyridine-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart