APO455827980 Apollo Scientific Ltd

3,4,6-Trichloropyridazine

  • MDL number MFCD00834957
  • Cas Number : 6082-66-2
  • Molecular Weight : 183.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-59
Synonym 3,4,6-Trichloro-1,2-diazine
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 96-97.5/2mm
Product Link https://store.apolloscientific.co.uk/product/346-trichloropyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/346-trichloropyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart