APO455827983 Apollo Scientific Ltd

2,8-Diazaspiro[4.5]decane, N2-BOC protected

  • MDL number MFCD06739056
  • Cas Number : 336191-17-4
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2,8-diazaspiro[4.5]decane-2-carboxylate
Inchi Code 1S/C13H24N2O2/c1-12(2,3)17-11(16)15-9-6-13(10-15)4-7-14-8-5-13/h14H,4-10H2,1-3H3
Inchi Key NFNCPNAVNRBDOU-UHFFFAOYSA-N

Description

Synonym tert-Butyl 2,8-diazaspiro[4.5]decane-2-carboxylate, 2-(tert-Butoxycarbonyl)-2,8-diazaspiro[4.5]decane
Storage temp. Store at 2-8
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/28-diazaspiro45decane-n2-boc-protected

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/28-diazaspiro45decane-n2-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart