APO455827984 Apollo Scientific Ltd

5-Amino-2-chloroisonicotinic acid

  • MDL number MFCD06659499
  • Cas Number : 58483-95-7
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-2-chloroisonicotinic acid
Inchi Code 1S/C6H5ClN2O2/c7-5-1-3(6(10)11)4(8)2-9-5/h1-2H,8H2,(H,10,11)
Inchi Key WCZUTMZMEAPPIX-UHFFFAOYSA-N

Description

Synonym 5-Amino-2-chloropyridine-4-carboxylic acid
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-chloroisonicotinic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-chloroisonicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart