APO455827986 Apollo Scientific Ltd

3-(Hydroxymethyl)-5-nitro-1H-indole, N-BOC protected

  • MDL number MFCD05864712
  • Cas Number : 914349-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)-5-nitro-1H-indole
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-hydroxymethyl-5-nitro-1h-indole-n-boc-protected

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-hydroxymethyl-5-nitro-1h-indole-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart