APO455827989 Apollo Scientific Ltd

6,8-Dibromoimidazo[1,2-a]pyrazine

  • MDL number MFCD08460056
  • Cas Number : 63744-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/68-dibromoimidazo12-apyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/68-dibromoimidazo12-apyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart