APO455827993 Apollo Scientific Ltd

2-Amino-4,5-dichlorophenol

  • MDL number MFCD12406838
  • Cas Number : 28443-57-4
  • Molecular Weight : 178.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-175
Synonym 4,5-Dichloro-2-hydroxyaniline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-45-dichlorophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-45-dichlorophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart