APO455827994 Apollo Scientific Ltd

2,6-Dibromobenzaldehyde

  • MDL number MFCD11040410
  • Cas Number : 67713-23-9
  • Molecular Weight : 263.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 89-91
Iupac Name 2,6-dibromobenzaldehyde
Inchi Code 1S/C7H4Br2O/c8-6-2-1-3-7(9)5(6)4-10/h1-4H
Inchi Key YDYNSAUGVGAOLO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 89-91
Product Link https://store.apolloscientific.co.uk/product/26-dibromobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dibromobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart