APO455828002 Apollo Scientific Ltd

3-Methyl-2-nitroaniline

  • MDL number MFCD00074851
  • Cas Number : 601-87-6
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 105
Iupac Name 3-methyl-2-nitroaniline
Inchi Code 1S/C7H8N2O2/c1-5-3-2-4-6(8)7(5)9(10)11/h2-4H,8H2,1H3
Inchi Key VDCZKCIEXGXCDJ-UHFFFAOYSA-N

Description

Synonym 3-Amino-2-nitrotoluene, 2-Amino-6-methylnitrobenzene, 2-Nitro-m-toluidine
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 105
Product Link https://store.apolloscientific.co.uk/product/3-methyl-2-nitroaniline

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/3-methyl-2-nitroaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart