APO455828004 Apollo Scientific Ltd

4-(Cyanomethyl)benzoic acid

  • MDL number MFCD00020382
  • Cas Number : 50685-26-2
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196
Synonym 4-Carboxyphenylacetonitrile, 4-Carboxybenzyl cyanide
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-cyanomethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-cyanomethylbenzoic-acid
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart