APO455828006 Apollo Scientific Ltd

2-(Bromomethyl)benzoic acid

  • MDL number MFCD00090420
  • Cas Number : 7115-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-150(dec.)
Synonym 2-Carboxybenzyl bromide, alpha-Bromo-o-toluic acid
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45789

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45789
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart