APO455828011 Apollo Scientific Ltd

5-Bromo-3-chloropyridine-2-carboxylic acid

  • MDL number MFCD12827552
  • Cas Number : 1189513-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-3-chloropicolinic acid, 5-Bromo-2-carboxy-3-chloropyridine
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46236

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46236
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart