APO455828014 Apollo Scientific Ltd

1-Methyl-6-phenyl-1H-imidazo[4,5-b]pyridin-2-amine

  • MDL number MFCD00210745
  • Cas Number : 105650-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-229
Synonym 2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine, 2-Amino-1-methyl-6-phenyl-4-azabenzimidazole
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12716

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12716
pictograms GHS06
hstatements H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart