APO455828019 Apollo Scientific Ltd

4-Bromoindan-2-one

  • MDL number MFCD09744095
  • Cas Number : 846032-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-75
Synonym 4-Bromo-1,3-dihydro-2H-inden-2-one, 4-Bromo-2-oxoindane, 4-Bromo-2,3-dihydro-2-oxo-1H-indene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46789

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=46789
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart