APO455828025 Apollo Scientific Ltd

2,4-Dibromo-1,3,5-trimethylbenzene

  • MDL number MFCD00013525
  • Cas Number : 6942-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-63
Synonym 2,4-Dibromomesitylene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 278-279
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59118

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=59118
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart