APO455828029 Apollo Scientific Ltd

3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one

  • MDL number MFCD09746273
  • Cas Number : 135352-71-5
  • Molecular Weight : 261.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-iodo-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one
Inchi Code 1S/C6H4IN3O/c7-3-1-8-5-4(3)6(11)10-2-9-5/h1-2H,(H2,8,9,10,11)
Inchi Key QHYIUMOXLCMESH-UHFFFAOYSA-N

Description

Synonym 3,7-Dihydro-5-iodo-4-oxo-4H-pyrrolo[2,3-d]pyrimidine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/37-dihydro-5-iodo-4h-pyrrolo23-dpyrimidin-4-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/37-dihydro-5-iodo-4h-pyrrolo23-dpyrimidin-4-one
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart