APO455828033 Apollo Scientific Ltd

2-Amino-3,7-dihydro-4H-pyrrolo[2,3-d]pyrimidin-4-one

  • MDL number MFCD09033259
  • Cas Number : 7355-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 323-324
Synonym 2-Amino-3H-pyrrolo[2,3-d]pyrimidin-4(7H)-one, 7-Deazaguanine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46809

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46809
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart