APO455828037 Apollo Scientific Ltd

Ethyl 5-nitro-1H-pyrrole-2-carboxylate

  • MDL number MFCD09025771
  • Cas Number : 36131-46-1
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146
Synonym 2-(Ethoxycarbonyl)-5-nitro-1H-pyrrole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-nitro-1h-pyrrole-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-nitro-1h-pyrrole-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart