APO455828040 Apollo Scientific Ltd

1H-Benzimidazole-4-carboxaldehyde

  • MDL number MFCD10698555
  • Cas Number : 144876-36-8
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-benzimidazole-4-carbaldehyde
Inchi Code 1S/C8H6N2O/c11-4-6-2-1-3-7-8(6)10-5-9-7/h1-5H,(H,9,10)
Inchi Key UNGJRTXGRUDJGM-UHFFFAOYSA-N

Description

Synonym 4-Formyl-1H-benzimidazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-benzimidazole-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-benzimidazole-4-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart