APO455828042 Apollo Scientific Ltd

4-Bromopyridine-2-carbonitrile

  • MDL number MFCD04065805
  • Cas Number : 62150-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-91
Synonym 4-Bromo-2-cyanopyridine, 4-Bromopicolinonitrile
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47119

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47119
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart