APO455828045 Apollo Scientific Ltd

5-Bromo-2-iodobenzyl bromide

  • MDL number MFCD12025360
  • Cas Number : 495414-06-7
  • Molecular Weight : 375.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 156
bp 83-85
Iupac Name 4-bromo-2-(bromomethyl)-1-iodobenzene
Inchi Code 1S/C7H5Br2I/c8-4-5-3-6(9)1-2-7(5)10/h1-3H,4H2
Inchi Key AHETZDTVQBIWSL-UHFFFAOYSA-N

Description

Melting Point 156
Synonym 4-Bromo-2-(bromomethyl)-1-iodobenzene, alpha,5-Dibromo-2-iodotoluene
Purity 95%
Flash Point 335/760mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 83-85
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-iodobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-iodobenzyl-bromide
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart