APO455828047 Apollo Scientific Ltd

Methyl 3-bromo-5-iodobenzoate

  • MDL number MFCD00673016
  • Cas Number : 188813-07-2
  • Molecular Weight : 340.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-63
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-5-iodobenzoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-5-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart