APO455828061 Apollo Scientific Ltd

5-(Benzyloxy)-3-iodo-1H-indole, N-BOC protected

  • MDL number MFCD05864771
  • Cas Number : 914349-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 5-(benzyloxy)-3-iodo-1H-indole-1-carboxylate, 5-(Benzyloxy)-1-(tert-butoxycarbonyl)-3-iodo-1H-indole
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29732

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29732
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart