APO455828062 Apollo Scientific Ltd

3-Bromo-1H-indole, N-BOC protected

  • MDL number MFCD05864772
  • Cas Number : 143259-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 3-bromo-1H-indole-1-carboxylate, 3-Bromo-1-(tert-butoxycarbonyl)-1H-indole
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-1h-indole-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-1h-indole-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart