APO455828075 Apollo Scientific Ltd

3-Bromo-7-methyl-1H-indole, N-BOC protected

  • MDL number MFCD05864788
  • Cas Number : 914349-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 3-bromo-7-methyl-1H-indole-1-carboxylate, 3-Bromo-1-(tert-butoxylcarbonyl)-7-methyl-1H-indole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29749

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29749
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart