APO455828079 Apollo Scientific Ltd

2-Amino-6-bromobenzonitrile

  • MDL number MFCD09834769
  • Cas Number : 77326-62-6
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 152-153
Iupac Name 2-amino-6-bromobenzonitrile
Inchi Code 1S/C7H5BrN2/c8-6-2-1-3-7(10)5(6)4-9/h1-3H,10H2
Inchi Key ZKZYTSGUONPOPF-UHFFFAOYSA-N

Description

Synonym 3-Bromo-2-cyanoaniline
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 152-153
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-bromobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-bromobenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart