APO455828083 Apollo Scientific Ltd

7,8-Dihydroquinolin-5(6H)-one

  • MDL number MFCD02181161
  • Cas Number : 53400-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Oxo-5,6,7,8-tetrahydroquinoline, 5,6,7,8-Tetrahydroquinolin-5-one
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46869

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46869
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart