APO455828084 Apollo Scientific Ltd

6-Chloro-4-cyclopropylquinazolin-2(1H)-one

  • MDL number MFCD11109434
  • Cas Number : 150878-36-7
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4-cyclopropyl-2(1H)-quinazolinone
Inchi Code 1S/C11H9ClN2O/c12-7-3-4-9-8(5-7)10(6-1-2-6)14-11(15)13-9/h3-5H,1-2H2,(H2,13,14,15)
Inchi Key XCLOLVRAVQUXNE-UHFFFAOYSA-N

Description

Synonym 6-Chloro-4-cyclopropyl-1,2-dihydro-2-oxoquinazoline
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-4-cyclopropylquinazolin-21h-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-4-cyclopropylquinazolin-21h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart