APO455828085 Apollo Scientific Ltd

2-Methyl-1,3-oxazole-4-carbohydrazide

  • MDL number MFCD11052651
  • Cas Number : 500341-65-1
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1,3-oxazole-4-carbohydrazide
Inchi Code 1S/C5H7N3O2/c1-3-7-4(2-10-3)5(9)8-6/h2H,6H2,1H3,(H,8,9)
Inchi Key GBQAWWYOPRFGMG-UHFFFAOYSA-N

Description

Synonym 2-Methyl-1,3-oxazole-4-carboxylic acid hydrazide, 4-(Hydrazinocarbonyl)-2-methyl-1,3-oxazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-13-oxazole-4-carbohydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-13-oxazole-4-carbohydrazide
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart