APO455828087 Apollo Scientific Ltd

2-Bromo-5-chloro-3-methylpyridine

  • MDL number MFCD03095062
  • Cas Number : 65550-77-8
  • Molecular Weight : 206.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 36-37
Iupac Name 2-bromo-5-chloro-3-methylpyridine
Inchi Code 1S/C6H5BrClN/c1-4-2-5(8)3-9-6(4)7/h2-3H,1H3
Inchi Key XEHWMXAUPXFOHZ-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-chloro-3-picoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 36-37
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-chloro-3-methylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-chloro-3-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart