APO455828088 Apollo Scientific Ltd

5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole

  • MDL number MFCD08449281
  • Cas Number : 1017788-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethylamine
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46975

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46975
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart