APO455828092 Apollo Scientific Ltd

2,3-Dichloro-5-(hydroxymethyl)pyridine

  • MDL number MFCD00671515
  • Cas Number : 54127-30-9
  • Molecular Weight : 178.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 76
Iupac Name (5,6-dichloro-3-pyridinyl)methanol
Inchi Code 1S/C6H5Cl2NO/c7-5-1-4(3-10)2-9-6(5)8/h1-2,10H,3H2
Inchi Key ZOFUUOULXZPZHP-UHFFFAOYSA-N

Description

Synonym (5,6-Dichloropyridin-3-yl)methanol
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 76
Product Link https://store.apolloscientific.co.uk/product/23-dichloro-5-hydroxymethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dichloro-5-hydroxymethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart