APO455828097 Apollo Scientific Ltd

6-Bromo-2-chloroquinoline

  • MDL number MFCD04115272
  • Cas Number : 1810-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Synonym 6-Bromo-2-chloro-1-azanaphthalene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-bromo-2-chloroquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-2-chloroquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart