APO455828101 Apollo Scientific Ltd

Quinoline-7-boronic acid

  • MDL number MFCD11100750
  • Cas Number : 629644-82-2
  • Molecular Weight : 172.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-quinolinylboronic acid
Inchi Code 1S/C9H8BNO2/c12-10(13)8-4-3-7-2-1-5-11-9(7)6-8/h1-6,12-13H
Inchi Key ZMEMBOIUHLWVRX-UHFFFAOYSA-N

Description

Synonym 7-Boronoquinoline
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/quinoline-7-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/quinoline-7-boronic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart