APO455828102 Apollo Scientific Ltd

Quinoline-7-carboxylic acid

  • MDL number MFCD00047617
  • Cas Number : 1078-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Carboxyquinoline, 7-Carboxy-1-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/quinoline-7-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/quinoline-7-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart