APO455828105 Apollo Scientific Ltd

2-Bromo-6-nitroanisole

  • MDL number MFCD09261208
  • Cas Number : 98775-19-0
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-65
Synonym 1-Bromo-2-methoxy-3-nitrobenzene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-nitroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart