APO455828109 Apollo Scientific Ltd

6-Iodoindoline

  • MDL number MFCD07371641
  • Cas Number : 115666-46-1
  • Molecular Weight : 245.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-iodoindoline
Inchi Code 1S/C8H8IN/c9-7-2-1-6-3-4-10-8(6)5-7/h1-2,5,10H,3-4H2
Inchi Key KGCBZEKELZUOFI-UHFFFAOYSA-N

Description

Synonym 2,3-Dihydro-6-iodo-1H-indole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-iodoindoline

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/6-iodoindoline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart