APO455828110 Apollo Scientific Ltd

4-(Bromomethyl)-5-methyl-1,3-dioxol-2-one

  • MDL number MFCD07787463
  • Cas Number : 80715-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-5-methyl-2-oxo-1,3-dioxole
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46927

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46927
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart